missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1R)-(-)-Camphorquinone 98.0+%, TCI America™
$110.11 - $436.69
Chemical Identifiers
| CAS | 10334-26-6 |
|---|---|
| Molecular Formula | C10H14O2 |
| Molecular Weight (g/mol) | 166.22 |
| MDL Number | MFCD00064160,MFCD00082863,MFCD00064160,MFCD00082863 |
| InChI Key | VNQXSTWCDUXYEZ-LDWIPMOCSA-N |
| Synonym | 1r,4s-1,7,7-trimethylnorbornane-2,3-dione, 1r,4r-4,7,7-trimethylbicyclo 2.2.1 heptane-2,3-dione, 1s,4r-4,7,7-trimethylbicyclo 2.2.1 heptane-2,3-dione |
| PubChem CID | 16211789 |
| IUPAC Name | (1R,4S)-1,7,7-trimethylbicyclo[2.2.1]heptane-2,3-dione |
| SMILES | CC1(C)[C@@H]2CC[C@@]1(C)C(=O)C2=O |
Chemical Identifiers
| 10334-26-6 | |
| 166.22 | |
| VNQXSTWCDUXYEZ-LDWIPMOCSA-N | |
| 16211789 | |
| CC1(C)[C@@H]2CC[C@@]1(C)C(=O)C2=O |
| C10H14O2 | |
| MFCD00064160,MFCD00082863,MFCD00064160,MFCD00082863 | |
| 1r,4s-1,7,7-trimethylnorbornane-2,3-dione, 1r,4r-4,7,7-trimethylbicyclo 2.2.1 heptane-2,3-dione, 1s,4r-4,7,7-trimethylbicyclo 2.2.1 heptane-2,3-dione | |
| (1R,4S)-1,7,7-trimethylbicyclo[2.2.1]heptane-2,3-dione |
Specifications
| 10334-26-6 | |
| Yellow | |
| MFCD00064160,MFCD00082863,MFCD00064160,MFCD00082863 | |
| VNQXSTWCDUXYEZ-LDWIPMOCSA-N | |
| (1R,4S)-1,7,7-trimethylbicyclo[2.2.1]heptane-2,3-dione | |
| 16211789 | |
| ≥98.0% (GC) | |
| (1R)-(-)-Camphorquinone |
| 200°C | |
| C10H14O2 | |
| 1r,4s-1,7,7-trimethylnorbornane-2,3-dione, 1r,4r-4,7,7-trimethylbicyclo 2.2.1 heptane-2,3-dione, 1s,4r-4,7,7-trimethylbicyclo 2.2.1 heptane-2,3-dione | |
| CC1(C)[C@@H]2CC[C@@]1(C)C(=O)C2=O | |
| 166.22 | |
| 166.22 | |
| Crystalline Powder |
Safety and Handling
TSCA : No