missing translation for 'onlineSavingsMsg'
Learn More

(1R)-(+)-Camphor, 98%

Catalog No. AAA1070836
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1070836 500 g
AAA1070814 25 g
AAA1070822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1070836 Supplier Thermo Scientific Chemicals Supplier No. A1070836
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 464-49-3 | C10H16O | 152.24 g/mol

(1R)-(+)-Camphor is used as a chiral intermediate and auxiliary. It is also used as a skin antipruritic and as an anti-infective agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1R)-(+)-Camphor is used as a chiral intermediate and auxiliary. It is also used as a skin antipruritic and as an anti-infective agent.

Solubility
Soluble in water (0.1 g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 464-49-3
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.24
MDL Number MFCD00064149,MFCD00074738,MFCD00064149,MFCD00064149
InChI Key DSSYKIVIOFKYAU-XVKPBYJWSA-N
Synonym +-camphor, d-camphor, 1r-+-camphor, r-camphor, +-bornan-2-one, r-+-camphor, 1r,4r-1,7,7-trimethylbicyclo 2.2.1 heptan-2-one, 1r,4r-camphor, camphor d, +-2-bornanone
PubChem CID 159055
ChEBI CHEBI:15396
IUPAC Name (1R,4R)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-one
SMILES CC1(C)[C@H]2CC[C@@]1(C)C(=O)C2

Specifications

Melting Point 175°C to 180°C
Density 0.99
Boiling Point 204°C
Flash Point 65°C (149°F)
Odor Camphor-like
Quantity 500 g
UN Number UN2717
Beilstein 2042745
Merck Index 14,1732
Solubility Information Soluble in water (0.1g/L at 20°C).
Optical Rotation +43° (c=10 in ethanol)
Formula Weight 152.24
Percent Purity 98%
Chemical Name or Material (1R)- (+)-Camphor
Show More Show Less
Hazard Category H228-H302-H315-H319-H335-H500
Hazard Statement GHS H Statement
H228-H315-H319
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P240-P241-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: CAMPHOR
EINECSNumber 207-355-2
RTECSNumber EX1260000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only