missing translation for 'onlineSavingsMsg'
Learn More

(1R,3S)-(+)-4-Cyclopentene-1,3-diol 1-acetate, 99%, stabilized

Catalog No. AC399340010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC399340010 1 g Glass bottle
AC399340050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC399340010 Supplier Thermo Scientific Chemicals Supplier No. 399340010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 60410-16-4 | C7H10O3 | 142.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 60410-16-4
Molecular Formula C7H10O3
Molecular Weight (g/mol) 142.15
MDL Number MFCD00078933
InChI Key IJDYOKVVRXZCFD-RQJHMYQMSA-N
Synonym 1s,4r-cis-4-acetoxy-2-cyclopenten-1-ol, 1r,4s-4-hydroxycyclopent-2-en-1-yl acetate, 1r,4s-4-hydroxycyclopent-2-enyl acetate, 1r,3s-+-cis-4-cyclopentene-1,3-diol 1-acetate, acetic acid 1r,4s-4-hydroxy-cyclopent-2-enyl ester, 1r,3s-+-4-cyclopentene-1,3-diol 1-acetate, 1s,4r-4-acetoxy-2-cyclopenten-1-ol, 1r,3s-4-cyclopentene-1,3-diol 1-acetate, 1r,4s-cis-4-hydroxy-2-cyclopentenyl acetate, 5beta-acetoxycyclopentene-3beta-ol
PubChem CID 10154103
IUPAC Name [(1R,4S)-4-hydroxycyclopent-2-en-1-yl] acetate
SMILES CC(=O)OC1CC(C=C1)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 43°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation +67° to +69° (20°C, 589nm) ((c=2.3 in chloroform))
Formula Weight 142.15
Percent Purity 99%
Physical Form Low Melting Solid
Chemical Name or Material (1R, 3S)-(+)-4-Cyclopentene-1, 3-diol 1-acetate, stabilzed
Show More Show Less

RUO – Research Use Only