accessibility menu, dialog, popup

UserName

CH3OOOH

CAS RN 60410-16-4

IUPAC Name: 4-hydroxycyclopent-2-en-1-yl acetate
Synonyms: 4-hydroxycyclopent-2-en-1-yl acetate
Molecular Weight (g/mol): 142.15
Molecular Formula: C7H10O3
InChi Key: IJDYOKVVRXZCFD-UHFFFAOYNA-N
SMILES: CC(=O)OC1CC(O)C=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results