missing translation for 'onlineSavingsMsg'
Learn More

(1R,3S)-3-(Carbobenzoxyamino)cyclohexanecarboxylic Acid 98.0+%, TCI America™

Catalog No. C2400100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2400100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C2400100MG Supplier TCI America Supplier No. C2400100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Reagent chemical suitable for laboratory applications

Chemical Identifiers

CAS 1259278-10-8
Molecular Formula C15H19NO4
Molecular Weight (g/mol) 277.32
InChI Key LTEORMGXUMWZCU-OLZOCXBDSA-N
Synonym (1R,3S)-3-(Cbz-amino)cyclohexanecarboxylic Acid
PubChem CID 23584443
IUPAC Name (1R,3S)-3-(phenylmethoxycarbonylamino)cyclohexane-1-carboxylic acid
SMILES C1CC(CC(C1)NC(=O)OCC2=CC=CC=C2)C(=O)O

Specifications

Color White
Quantity 100 mg
Formula Weight 277.32
Percent Purity ≥98.0% (HPLC,T)
Grade Research
Physical Form Crystalline Powder
Chemical Name or Material (1R,3S)-3-(Carbobenzoxyamino)cyclohexanecarboxylic Acid