CAS RN 1259278-10-8
IUPAC Name:
3-{[(benzyloxy)carbonyl]amino}cyclohexane-1-carboxylic acid
Synonyms:
3-{[(benzyloxy)carbonyl]amino}cyclohexane-1-carboxylic acid
Molecular Weight (g/mol):
277.32
Molecular Formula:
C15H19NO4
InChi Key:
LTEORMGXUMWZCU-UHFFFAOYNA-N
SMILES:
OC(=O)C1CCCC(C1)NC(=O)OCC1=CC=CC=C1
1
–
1
of
1
results
1
(1R,3S)-3-(Carbobenzoxyamino)cyclohexanecarboxylic Acid 98.0+%, TCI America™
CAS: 1259278-10-8 Molecular Formula: C15H19NO4 Molecular Weight (g/mol): 277.32 InChI Key: LTEORMGXUMWZCU-OLZOCXBDSA-N Synonym: (1R,3S)-3-(Cbz-amino)cyclohexanecarboxylic Acid PubChem CID: 23584443 IUPAC Name: (1R,3S)-3-(phenylmethoxycarbonylamino)cyclohexane-1-carboxylic acid SMILES: C1CC(CC(C1)NC(=O)OCC2=CC=CC=C2)C(=O)O