missing translation for 'onlineSavingsMsg'
Learn More

(1R,3S)-3-Aminocyclopentanecarboxylic acid, 95%, 98% ee

Catalog No. p-4510676
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC362270010 1 g Glass Bottle
AC362270050 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC362270010 Supplier Thermo Scientific Chemicals Supplier No. 362270010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 71830-08-5 | C6H11NO2 | 129.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 71830-08-5
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.16
InChI Key MLLSSTJTARJLHK-UHNVWZDZSA-N
Synonym 1r,3s-3-aminocyclopentanecarboxylic acid, 1r,3s-3-aminocyclopentane-1-carboxylic acid, --1r,3s-3-aminocyclopentanecarboxylic acid, cis-3-aminocyclopentanecarboxylic acid, cyclopentanecarboxylic acid, 3-amino-, 1r,3s, 1r,3s---3-aminocyclopentanecarboxylic acid, cis-3-amino-cyclopentanecarboxylic acid, 1r,3s-3-amino-cyclopentanecarboxylic acid, pubchem18731, 1r,3s---3-amino-cyclopentanecarboxylic acid
PubChem CID 1502035
IUPAC Name (1R,3S)-3-aminocyclopentane-1-carboxylic acid
SMILES C1CC(CC1C(=O)O)N

Specifications

Melting Point 172.1°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 129.16
Percent Purity 95%,98% ee
Physical Form Crystalline Powder
Chemical Name or Material (1R, 3S)-3-Aminocyclopentanecarboxylic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor

RUO – Research Use Only