accessibility menu, dialog, popup

UserName

CAS RN 71830-08-5

IUPAC Name: (1R,3S)-3-azaniumylcyclopentane-1-carboxylate
Synonyms: (1R,3S)-3-ammoniocyclopentane-1-carboxylate
Molecular Weight (g/mol): 129.16
Molecular Formula: C6H11NO2
InChi Key: MLLSSTJTARJLHK-UHNVWZDZSA-N
SMILES: [NH3+][C@H]1CC[C@H](C1)C([O-])=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)