missing translation for 'onlineSavingsMsg'
Learn More

(1R,3S)-3-Aminocyclohexanecarboxylic Acid 98.0+%, TCI America™

Catalog No. a2287200mg
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2287200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A2287200MG Supplier TCI America Supplier No. A2287200MG

Chemical Identifiers

CAS 81131-39-7
Molecular Formula C7H13NO2
Molecular Weight (g/mol) 143.186
MDL Number MFCD09753647
InChI Key CKTUXQBZPWBFDX-RITPCOANSA-N
PubChem CID 3082488
IUPAC Name (1R,3S)-3-aminocyclohexane-1-carboxylic acid
SMILES C1CC(CC(C1)N)C(=O)O

Specifications

Melting Point 279°C
Color White
Quantity 200 mg
Formula Weight 143.19
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (1R,3S)-3-Aminocyclohexanecarboxylic Acid