accessibility menu, dialog, popup

UserName

CAS RN 81131-39-7

NH2OHO

CAS RN 81131-39-7

IUPAC Name: 3-aminocyclohexane-1-carboxylic acid
Synonyms: 3-aminocyclohexane-1-carboxylic acid
Molecular Weight (g/mol): 143.19
Molecular Formula: C7H13NO2
InChi Key: CKTUXQBZPWBFDX-UHFFFAOYNA-N
SMILES: NC1CCCC(C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results