missing translation for 'onlineSavingsMsg'
Learn More

(1R,2S)-(+)-cis-1-Amino-2-indanol, 98%

Catalog No. AC297530100
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC297530100 10 g Glass bottle
AC297530010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC297530100 Supplier Thermo Scientific Chemicals Supplier No. 297530100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 136030-00-7 | C9H12NO | 150.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 136030-00-7
Molecular Formula C9H12NO
Molecular Weight (g/mol) 150.20
MDL Number MFCD00216656,MFCD01318239
InChI Key LOPKSXMQWBYUOI-DTWKUNHWSA-O
Synonym 1r,2s-1-amino-2-indanol, 1r,2s-1-amino-2,3-dihydro-1h-inden-2-ol, cis-1-amino-2-indanol, 1r,2s-+-cis-1-amino-2-indanol, cis-1-amino-2,3-dihydro-1h-inden-2-ol, 1r,2s-+-1-amino-2-hydroxyindan, 1r,2s-1-amino-2-hydroxyindane, 1r,2s-+-1-amino-2-indanol, 1r,2s-+-cis-1-aminoindan-2-ol, 1r,2s-cis-1-amino-2-indanol
PubChem CID 2725045
IUPAC Name (1R,2S)-2-hydroxy-2,3-dihydro-1H-inden-1-aminium
SMILES [NH3+][C@H]1[C@@H](O)CC2=CC=CC=C12

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 117°C to 121°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 10 g
Specific Rotation Condition +44.5° (20°C c=1,methanol)
Specific Rotation + 44.5
Solubility Information Solubility in water: soluble
Formula Weight 149.19
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (1R, 2S)-(+)-cis-1-Amino-2-indanol, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

RUO – Research Use Only