missing translation for 'onlineSavingsMsg'
Learn More

(1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl 2,2-Dihydroxyacetate 98.0+%, TCI America™

Catalog No. I079025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I079025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I079025G Supplier TCI America Supplier No. I079025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 111969-64-3
Molecular Formula C12H22O4
Molecular Weight (g/mol) 230.304
MDL Number MFCD00792503
InChI Key BWZMJRSMHQDFIT-KXUCPTDWSA-N
Synonym 1r,2s,5r-2-isopropyl-5-methylcyclohexyl 2,2-dihydroxyacetate, l-menthyl glyoxylate hydrate, 1r,2s,5r-5-methyl-2-1-methylethyl cyclohexyl dihydroxy-acetate, l-menthyl glyoxylate, l-menthylglyoxylate hydrate, --menthyl 2,2-dihydroxyacetate, 1r,2s,5r-5-methyl-2-1-methylethyl cyclohexyl di, 2,2-dihydroxyacetic acid 1r,3r,4s-p-menth-3-yl ester, 1r,2s,5r-5-methyl-2-propan-2-ylcyclohexyl 2,2-dihydroxyacetate, 2,2-dihydroxyacetic acid 1r,2s,5r-2-isopropyl-5-methylcyclohexyl ester
PubChem CID 7373179
IUPAC Name [(1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexyl] 2,2-dihydroxyacetate
SMILES CC1CCC(C(C1)OC(=O)C(O)O)C(C)C

Specifications

Melting Point 81°C
Color White
Quantity 25 g
UN Number 3077
Formula Weight 230.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl 2,2-Dihydroxyacetate