missing translation for 'onlineSavingsMsg'
Learn More

(1R,2S,3R,4R)-2,3-Dihydroxy-4-(hydroxymethyl)-1-aminocyclopentane hydrochloride, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 362522500

Quantity 250mg
Packaging Glass bottle

 View more versions of this product

Catalog No. AC362522500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

79200-57-0
183.63
1r,2s,3r,5r-3-amino-5-hydroxymethyl cyclopentane-1,2-diol hydrochloride, 1r,2s,3r,4r-2,3-dihydroxy-4-hydroxymethyl-1-aminocyclopentane hydrochloride, n-4-mesityl-1,3-thiazol-2-yl benzamide, 1r,2s,3r,5r-3-amino-5-hydroxymethyl cyclopentane-1,2-diol hcl, 1r,2s,3r,4r-2,3-dihydroxy-4-hydroxymethyl-1-am, 1r,2s,3r,4r-2,3-dihydroxy-4-hydroxymethyl cyclopentanaminium chloride, 1r,2s,3r,4r-2,3-dihydroxy-4-hydroxylmethyl-1-aminocyclopentan hydro-chloride, 1r,2s,3r,5r-3-amino-5-hydroxymethyl-cyclopentane-1,2-diol hydrochloride
(1R,2S,3R,5R)-3-amino-5-(hydroxymethyl)cyclopentane-1,2-diol;hydrochloride
C6H13NO3·HCl
BLTXEPQZAMUGID-ILLHIODVSA-N
2734536
C1C(C(C(C1N)O)O)CO.Cl

Specifications

(1R,2S,3R,4R)-2,3-Dihydroxy-4-(hydroxymethyl)-1-aminocyclopentane hydrochloride
94% min. (HPLC)
C6H13NO3·HCl
1r,2s,3r,5r-3-amino-5-hydroxymethyl cyclopentane-1,2-diol hydrochloride, 1r,2s,3r,4r-2,3-dihydroxy-4-hydroxymethyl-1-aminocyclopentane hydrochloride, n-4-mesityl-1,3-thiazol-2-yl benzamide, 1r,2s,3r,5r-3-amino-5-hydroxymethyl cyclopentane-1,2-diol hcl, 1r,2s,3r,4r-2,3-dihydroxy-4-hydroxymethyl-1-am, 1r,2s,3r,4r-2,3-dihydroxy-4-hydroxymethyl cyclopentanaminium chloride, 1r,2s,3r,4r-2,3-dihydroxy-4-hydroxylmethyl-1-aminocyclopentan hydro-chloride, 1r,2s,3r,5r-3-amino-5-hydroxymethyl-cyclopentane-1,2-diol hydrochloride
C1C(C(C(C1N)O)O)CO.Cl
183.63
183.63
79200-57-0
Glass bottle
250mg
BLTXEPQZAMUGID-ILLHIODVSA-N
(1R,2S,3R,5R)-3-amino-5-(hydroxymethyl)cyclopentane-1,2-diol;hydrochloride
2734536
95%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions

Promo
Expires: 06-30-2027
Get Up to $1,800 in Chemicals with Purchase
Spend $250 or more on Thermo Fisher Scientific chemicals and get up to $1,800 to spend on additional chemicals.

Adding...