Learn More
(1R,2S,3R,4R)-2,3-Dihydroxy-4-(hydroxymethyl)-1-aminocyclopentane hydrochloride, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 362520010
| Quantity | 1g |
|---|---|
| Packaging | Glass bottle |
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 79200-57-0 | |
| 183.63 | |
| 1r,2s,3r,5r-3-amino-5-hydroxymethyl cyclopentane-1,2-diol hydrochloride, 1r,2s,3r,4r-2,3-dihydroxy-4-hydroxymethyl-1-aminocyclopentane hydrochloride, n-4-mesityl-1,3-thiazol-2-yl benzamide, 1r,2s,3r,5r-3-amino-5-hydroxymethyl cyclopentane-1,2-diol hcl, 1r,2s,3r,4r-2,3-dihydroxy-4-hydroxymethyl-1-am, 1r,2s,3r,4r-2,3-dihydroxy-4-hydroxymethyl cyclopentanaminium chloride, 1r,2s,3r,4r-2,3-dihydroxy-4-hydroxylmethyl-1-aminocyclopentan hydro-chloride, 1r,2s,3r,5r-3-amino-5-hydroxymethyl-cyclopentane-1,2-diol hydrochloride | |
| (1R,2S,3R,5R)-3-amino-5-(hydroxymethyl)cyclopentane-1,2-diol;hydrochloride |
| C6H13NO3·HCl | |
| BLTXEPQZAMUGID-ILLHIODVSA-N | |
| 2734536 | |
| C1C(C(C(C1N)O)O)CO.Cl |
Specifications
| (1R,2S,3R,4R)-2,3-Dihydroxy-4-(hydroxymethyl)-1-aminocyclopentane hydrochloride | |
| 94% min. (HPLC) | |
| C6H13NO3·HCl | |
| 1r,2s,3r,5r-3-amino-5-hydroxymethyl cyclopentane-1,2-diol hydrochloride, 1r,2s,3r,4r-2,3-dihydroxy-4-hydroxymethyl-1-aminocyclopentane hydrochloride, n-4-mesityl-1,3-thiazol-2-yl benzamide, 1r,2s,3r,5r-3-amino-5-hydroxymethyl cyclopentane-1,2-diol hcl, 1r,2s,3r,4r-2,3-dihydroxy-4-hydroxymethyl-1-am, 1r,2s,3r,4r-2,3-dihydroxy-4-hydroxymethyl cyclopentanaminium chloride, 1r,2s,3r,4r-2,3-dihydroxy-4-hydroxylmethyl-1-aminocyclopentan hydro-chloride, 1r,2s,3r,5r-3-amino-5-hydroxymethyl-cyclopentane-1,2-diol hydrochloride | |
| C1C(C(C(C1N)O)O)CO.Cl | |
| 183.63 | |
| 183.63 |
| 79200-57-0 | |
| Glass bottle | |
| 1g | |
| BLTXEPQZAMUGID-ILLHIODVSA-N | |
| (1R,2S,3R,5R)-3-amino-5-(hydroxymethyl)cyclopentane-1,2-diol;hydrochloride | |
| 2734536 | |
| 95% |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
Promotions