missing translation for 'onlineSavingsMsg'
Learn More

(1R,2S)-2-[N-Benzyl-N-(mesitylenesulfonyl)amino]-1-phenyl-1-propanol 98.0+%, TCI America™

Catalog No. B21035G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21035G 5 g
B21031G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21035G Supplier TCI America Supplier No. B21035G

Chemical Identifiers

CAS 187324-63-6
Molecular Formula C25H29NO3S
Molecular Weight (g/mol) 423.571
MDL Number MFCD02093446
InChI Key ZPERKXJNXPLVAC-URXFXBBRSA-N
Synonym (1R,2S)-N-Benzyl-N-(mesitylenesulfonyl)norephedrine, (1R,2S)-2-[N-Benzyl-N-(2,4,6-trimethylbenzenesulfonyl)amino]-1-phenyl-1-propanol
PubChem CID 11166154
IUPAC Name N-benzyl-N-[(1R,2S)-1-hydroxy-1-phenylpropan-2-yl]-2,4,6-trimethylbenzenesulfonamide
SMILES CC1=CC(=C(C(=C1)C)S(=O)(=O)N(CC2=CC=CC=C2)C(C)C(C3=CC=CC=C3)O)C

Specifications

Melting Point 132°C
Color White-Yellow
Quantity 5 g
Formula Weight 423.57
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material (1R,2S)-2-[N-Benzyl-N-(mesitylenesulfonyl)amino]-1-phenyl-1-propanol