missing translation for 'onlineSavingsMsg'
Learn More

(1R,2S)-(+)-1-Amino-2-indanol 98.0+%, TCI America™

Catalog No. A16231G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A16231G 1 g
A16235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A16231G Supplier TCI America Supplier No. A16231G

Chemical Identifiers

CAS 136030-00-7
Molecular Formula C9H12NO
Molecular Weight (g/mol) 150.20
MDL Number MFCD00216656,MFCD01318239
InChI Key LOPKSXMQWBYUOI-DTWKUNHWSA-O
Synonym 1r,2s-1-amino-2-indanol, 1r,2s-1-amino-2,3-dihydro-1h-inden-2-ol, cis-1-amino-2-indanol, 1r,2s-+-cis-1-amino-2-indanol, cis-1-amino-2,3-dihydro-1h-inden-2-ol, 1r,2s-+-1-amino-2-hydroxyindan, 1r,2s-1-amino-2-hydroxyindane, 1r,2s-+-1-amino-2-indanol, 1r,2s-+-cis-1-aminoindan-2-ol, 1r,2s-cis-1-amino-2-indanol
PubChem CID 2725045
IUPAC Name (1R,2S)-2-hydroxy-2,3-dihydro-1H-inden-1-aminium
SMILES [NH3+][C@H]1[C@@H](O)CC2=CC=CC=C12

Specifications

Melting Point 116°C
Color White-Yellow
Quantity 1 g
Formula Weight 149.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (1R,2S)-(+)-1-Amino-2-indanol
EINECSNumber (4)-1717
TSCA No