missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1R,2R,3S,5R)-(-)-2,3-Pinanediol 98.0+%, TCI America™
$64.56 - $206.95
Chemical Identifiers
| CAS | 22422-34-0 |
|---|---|
| Molecular Formula | C10H18O2 |
| Molecular Weight (g/mol) | 170.25 |
| MDL Number | MFCD09955216 |
| InChI Key | MOILFCKRQFQVFS-UHFFFAOYNA-N |
| Synonym | 1r,2r,3s,5r---2,3-pinanediol, 1r,2r,3s,5r---pinanediol, 1r,2r,3s,5r-2,6,6-trimethylbicyclo 3.1.1 heptane-2,3-diol, --2,3-pinanediol, unii-r58l0w3a75, --cis-pinane-cis-2,3-diol, pinanediol, --2-hydroxyisopinocampheol, bicyclo 3.1.1 heptane-2,3-diol, 2,6,6-trimethyl-, 1r,2r,3s,5r |
| PubChem CID | 6553875 |
| IUPAC Name | 2,6,6-trimethylbicyclo[3.1.1]heptane-2,3-diol |
| SMILES | CC1(C)C2CC1C(C)(O)C(O)C2 |
Chemical Identifiers
| 22422-34-0 | |
| 170.25 | |
| MOILFCKRQFQVFS-UHFFFAOYNA-N | |
| 6553875 | |
| CC1(C)C2CC1C(C)(O)C(O)C2 |
| C10H18O2 | |
| MFCD09955216 | |
| 1r,2r,3s,5r---2,3-pinanediol, 1r,2r,3s,5r---pinanediol, 1r,2r,3s,5r-2,6,6-trimethylbicyclo 3.1.1 heptane-2,3-diol, --2,3-pinanediol, unii-r58l0w3a75, --cis-pinane-cis-2,3-diol, pinanediol, --2-hydroxyisopinocampheol, bicyclo 3.1.1 heptane-2,3-diol, 2,6,6-trimethyl-, 1r,2r,3s,5r | |
| 2,6,6-trimethylbicyclo[3.1.1]heptane-2,3-diol |
Specifications
| 22422-34-0 | |
| White-Yellow | |
| C10H18O2 | |
| 1r,2r,3s,5r---2,3-pinanediol, 1r,2r,3s,5r---pinanediol, 1r,2r,3s,5r-2,6,6-trimethylbicyclo 3.1.1 heptane-2,3-diol, --2,3-pinanediol, unii-r58l0w3a75, --cis-pinane-cis-2,3-diol, pinanediol, --2-hydroxyisopinocampheol, bicyclo 3.1.1 heptane-2,3-diol, 2,6,6-trimethyl-, 1r,2r,3s,5r | |
| CC1(C)C2CC1C(C)(O)C(O)C2 | |
| 170.25 | |
| 170.25 | |
| Crystalline Powder |
| 56°C | |
| 90°C | |
| MFCD09955216 | |
| MOILFCKRQFQVFS-UHFFFAOYNA-N | |
| 2,6,6-trimethylbicyclo[3.1.1]heptane-2,3-diol | |
| 6553875 | |
| ≥98.0% (GC) | |
| (1R,2R,3S,5R)-(-)-2,3-Pinanediol |
Safety and Handling
TSCA : No