missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R,3S,5R)-(-)-2,3-Pinanediol 98.0+%, TCI America™

Catalog No. P22451G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P22451G 1 g
P22455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P22451G Supplier TCI America Supplier No. P22451G

Chemical Identifiers

CAS 22422-34-0
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.25
MDL Number MFCD09955216
InChI Key MOILFCKRQFQVFS-UHFFFAOYNA-N
Synonym 1r,2r,3s,5r---2,3-pinanediol, 1r,2r,3s,5r---pinanediol, 1r,2r,3s,5r-2,6,6-trimethylbicyclo 3.1.1 heptane-2,3-diol, --2,3-pinanediol, unii-r58l0w3a75, --cis-pinane-cis-2,3-diol, pinanediol, --2-hydroxyisopinocampheol, bicyclo 3.1.1 heptane-2,3-diol, 2,6,6-trimethyl-, 1r,2r,3s,5r
PubChem CID 6553875
IUPAC Name 2,6,6-trimethylbicyclo[3.1.1]heptane-2,3-diol
SMILES CC1(C)C2CC1C(C)(O)C(O)C2

Specifications

Melting Point 56°C
Color White-Yellow
Boiling Point 90°C
Quantity 1 g
Formula Weight 170.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (1R,2R,3S,5R)-(-)-2,3-Pinanediol