Learn More
(1R,2R)-(-)-2-(Diphenylphosphino)cyclohexylamine, 97+%
CAS: 452304-59-5 | C18H22NP | 283.36 g/mol
Supplier: Thermo Scientific Chemicals H60084MD
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilityInsoluble in water.
Notes
Air sensitive. Keep away from strong oxidizing agents.
Specifications
| (1R,2R)-(-)-2-(Diphenylphosphino)cyclohexylamine | |
| 43°C to 48°C | |
| C18H22NP | |
| 250 mg | |
| 1r,2r-2-diphenylphosphino cyclohexanamine, 1r,2r-2-diphenylphosphanyl cyclohexan-1-amine, 1r,2r---2-diphenylphosphino cyclohexylamine, 1r,2r-2-diphenylphosphino cyclohexylamine, 1r-2alpha-diphenylphosphino cyclohexane-1beta-amine, 1r,2r---2-diphenylphosphino cyclohexylamine, 97+%, 1r,2r-2-diphenylphosphino-1-aminocyclohexane, 1r,2r-2-diphenylphosphino-1-aminocyclohexane,min | |
| ZATLZEHZPXYMFE-YTXKPMTANA-N | |
| N[C@@H]1CCCC[C@H]1P(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 283.36 | |
| 283.35 |
| 452304-59-5 | |
| >110°C (230°F) | |
| MFCD17013996 | |
| Air Sensitive | |
| Insoluble in water. | |
| −79° (c=0.5 in Chloroform) | |
| (1R,2R)-2-diphenylphosphanylcyclohexan-1-amine | |
| 11166387 | |
| ≥97% |
Chemical Identifiers
| 452304-59-5 | |
| 283.36 | |
| ZATLZEHZPXYMFE-YTXKPMTANA-N | |
| 11166387 |
| C18H22NP | |
| MFCD17013996 | |
| 1r,2r-2-diphenylphosphino cyclohexanamine, 1r,2r-2-diphenylphosphanyl cyclohexan-1-amine, 1r,2r---2-diphenylphosphino cyclohexylamine, 1r,2r-2-diphenylphosphino cyclohexylamine, 1r-2alpha-diphenylphosphino cyclohexane-1beta-amine, 1r,2r---2-diphenylphosphino cyclohexylamine, 97+%, 1r,2r-2-diphenylphosphino-1-aminocyclohexane, 1r,2r-2-diphenylphosphino-1-aminocyclohexane,min | |
| N[C@@H]1CCCC[C@H]1P(C1=CC=CC=C1)C1=CC=CC=C1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only