accessibility menu, dialog, popup

UserName

CAS RN 452304-59-5

IUPAC Name: (1R,2R)-2-(diphenylphosphanyl)cyclohexan-1-amine
Synonyms: (1R,2R)-2-(diphenylphosphanyl)cyclohexan-1-amine
Molecular Weight (g/mol): 283.36
Molecular Formula: C18H22NP
InChi Key: ZATLZEHZPXYMFE-YTXKPMTANA-N
SMILES: N[C@@H]1CCCC[C@H]1P(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results