missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R)-2-(Anthracene-2,3-dicarboximido)cyclohexanecarboxylic Acid 98.0+%, TCI America™

Catalog No. a1657100mg
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1657100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A1657100MG Supplier TCI America Supplier No. A1657100MG

Chemical Identifiers

CAS 446044-44-6
Molecular Formula C23H19NO4
Molecular Weight (g/mol) 373.408
InChI Key IIJKRPVAVXANME-YLJYHZDGSA-N
Synonym N-[(1R,2R)-2-Carboxycyclohexyl]anthracene-2,3-dicarboximide
PubChem CID 44607940
IUPAC Name (1R,2R)-2-(1,3-dioxonaphtho[2,3-f]isoindol-2-yl)cyclohexane-1-carboxylic acid
SMILES C1CCC(C(C1)C(=O)O)N2C(=O)C3=CC4=CC5=CC=CC=C5C=C4C=C3C2=O

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 373.41
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (1R,2R)-2-(Anthracene-2,3-dicarboximido)cyclohexanecarboxylic Acid