Learn More
(1R,2R)-2-Aminocyclohexanecarboxylic acid, 97%
CAS: 26685-83-6 | C7H13NO2 | 143.186 g/mol
Supplier: Thermo Scientific Chemicals H5241803
Description
(1R,2R)-2-Aminocyclohexanecarboxylic acid is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(1R,2R)-2-Aminocyclohexanecarboxylic acid is used as an organic chemical synthesis intermediate.
Solubility
Soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
| (1R,2R)-2-Aminocyclohexanecarboxylic acid | |
| C7H13NO2 | |
| 1r,2r-2-aminocyclohexanecarboxylic acid, trans-2-amino-1-cyclohexanecarboxylic acid, trans-2-aminocyclohexanecarboxylic acid, 1r,2r-2-aminocyclohexane-1-carboxylic acid, trans-2-amino-cyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 2-amino-, trans, cyclohexanecarboxylic acid, 2-amino-, 1r,2r, cyclohexanecarboxylic acid, 2-amino-, 1r,2r-rel, 2-nchca, 2-aminocyclohexanecarboxylic acid # | |
| USQHEVWOPJDAAX-PHDIDXHHSA-N | |
| (1R,2R)-2-aminocyclohexane-1-carboxylic acid | |
| 143.186 | |
| −77° (c=1 in Water) | |
| 97% |
| 26685-83-6 | |
| MFCD08276263 | |
| Soluble in water. | |
| C1CCC(C(C1)C(=O)O)N | |
| 1 g | |
| 2724640 | |
| 143.19 |
Chemical Identifiers
| 26685-83-6 | |
| 143.186 | |
| USQHEVWOPJDAAX-PHDIDXHHSA-N | |
| 2724640 | |
| C1CCC(C(C1)C(=O)O)N |
| C7H13NO2 | |
| MFCD08276263 | |
| 1r,2r-2-aminocyclohexanecarboxylic acid, trans-2-amino-1-cyclohexanecarboxylic acid, trans-2-aminocyclohexanecarboxylic acid, 1r,2r-2-aminocyclohexane-1-carboxylic acid, trans-2-amino-cyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 2-amino-, trans, cyclohexanecarboxylic acid, 2-amino-, 1r,2r, cyclohexanecarboxylic acid, 2-amino-, 1r,2r-rel, 2-nchca, 2-aminocyclohexanecarboxylic acid # | |
| (1R,2R)-2-aminocyclohexane-1-carboxylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only