missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R)-2-Aminocyclohexanecarboxylic acid, 97%

Catalog No. p-7034720
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52418MD 250 mg
AAH5241803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52418MD Supplier Thermo Scientific Chemicals Supplier No. H52418MD

missing translation for 'validMainframeMsg'

CAS: 26685-83-6 | C7H13NO2 | 143.186 g/mol

(1R,2R)-2-Aminocyclohexanecarboxylic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1R,2R)-2-Aminocyclohexanecarboxylic acid is used as an organic chemical synthesis intermediate.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 26685-83-6
Molecular Formula C7H13NO2
Molecular Weight (g/mol) 143.186
MDL Number MFCD08276263
InChI Key USQHEVWOPJDAAX-PHDIDXHHSA-N
Synonym 1r,2r-2-aminocyclohexanecarboxylic acid, trans-2-amino-1-cyclohexanecarboxylic acid, trans-2-aminocyclohexanecarboxylic acid, 1r,2r-2-aminocyclohexane-1-carboxylic acid, trans-2-amino-cyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 2-amino-, trans, cyclohexanecarboxylic acid, 2-amino-, 1r,2r, cyclohexanecarboxylic acid, 2-amino-, 1r,2r-rel, 2-nchca, 2-aminocyclohexanecarboxylic acid #
PubChem CID 2724640
IUPAC Name (1R,2R)-2-aminocyclohexane-1-carboxylic acid
SMILES C1CCC(C(C1)C(=O)O)N

Specifications

Solubility Information Soluble in water.
Quantity 250 mg
Optical Rotation −77° (c=1 in Water)
Formula Weight 143.19
Percent Purity 97%
Chemical Name or Material (1R,2R)-2-Aminocyclohexanecarboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only