missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R)-(+)-1,2-Diaminocyclohexane L-tartrate, 98%

Catalog No. AAH6017204
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6017204 2 g
AAH6017209 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6017204 Supplier Thermo Scientific Chemicals Supplier No. H6017204
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 39961-95-0 | C10H20N2O6 | 264.278 g/mol

(1R,2R)-(+)-1,2-Diaminocyclohexane L-tartrate is a useful chemical for the synthesis of chiral ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1R,2R)-(+)-1,2-Diaminocyclohexane L-tartrate is a useful chemical for the synthesis of chiral ligands.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 39961-95-0
Molecular Formula C10H20N2O6
Molecular Weight (g/mol) 264.278
MDL Number MFCD00191979
InChI Key GDOTUTAQOJUZOF-ZXZVGZDWSA-N
Synonym 1r,2r-+-1,2-diaminocyclohexane l-tartrate, 1r,2r-cyclohexane-1,2-diamine 2r,3r-2,3-dihydroxysuccinate, 1r,2r-+-1,2-cyclohexanediamine l-tartrate, 1r,2r---cyclohexane-1,2-diamine l-tartrate salt, 1r-trans-1,2-diaminocyclohexane l-tartrate, 1r,2r-+-cyclohexane-1,2-diamine l-tartrate, 1r,2r-1,2-diaminocyclohexane l-tartrate, 1r,2r-cyclohexane-1,2-diamine; l +-tartaric acid, pubchem17364, ksc221k4n
PubChem CID 11448443
IUPAC Name (1R,2R)-cyclohexane-1,2-diamine;(2R,3R)-2,3-dihydroxybutanedioic acid
SMILES C1CCC(C(C1)N)N.C(C(C(=O)O)O)(C(=O)O)O

Specifications

Melting Point 273°C (decomposition)
Quantity 2 g
Optical Rotation +12° (c=4 in Water)
Formula Weight 264.28
Percent Purity 99%
Chemical Name or Material (1R,2R)-(+)-1,2-Diaminocyclohexane L-tartrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only