missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R)-1,2-Cyclohexanedimethanol 98.0+%, TCI America™

Catalog No. C29785G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29785G 5 g
C29781G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C29785G Supplier TCI America Supplier No. C29785G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 65376-05-8
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.214
MDL Number MFCD16660860
InChI Key XDODWINGEHBYRT-YUMQZZPRSA-N
Synonym (1R,2R)-1,2-Bis(hydroxymethyl)cyclohexane
PubChem CID 11217323
IUPAC Name [(1R,2R)-2-(hydroxymethyl)cyclohexyl]methanol
SMILES C1CCC(C(C1)CO)CO

Specifications

Melting Point 65°C
Color White
Boiling Point 113°C
Quantity 5 g
Formula Weight 144.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (1R,2R)-1,2-Cyclohexanedimethanol