accessibility menu, dialog, popup

UserName

OHHO

CAS RN 65376-05-8

IUPAC Name: [2-(hydroxymethyl)cyclohexyl]methanol
Synonyms: [2-(hydroxymethyl)cyclohexyl]methanol
Molecular Weight (g/mol): 144.21
Molecular Formula: C8H16O2
InChi Key: XDODWINGEHBYRT-UHFFFAOYNA-N
SMILES: OCC1CCCCC1CO
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results