missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R)-1,2-Bis(2-hydroxyphenyl)ethylenediamine 95.0+%, TCI America™

Catalog No. B3012100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3012100MG 100 mg
B30121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B3012100MG Supplier TCI America Supplier No. B3012100MG
Only null left

Chemical Identifiers

CAS 870991-70-1
Molecular Formula C14H16N2O2
Molecular Weight (g/mol) 244.294
MDL Number MFCD09751760
InChI Key MRNPLGLZBUDMRE-ZIAGYGMSSA-N
PubChem CID 16218403
IUPAC Name 2-[(1R,2R)-1,2-diamino-2-(2-hydroxyphenyl)ethyl]phenol
SMILES C1=CC=C(C(=C1)C(C(C2=CC=CC=C2O)N)N)O

Specifications

Melting Point 158°C
Quantity 100 mg
Synonym (1R,2R)-1,2-Diamino-1,2-bis(2-hydroxyphenyl)ethane, 2,2′-[(1R,2R)-1,2-Diaminoethylene]bisphenol
Formula Weight 244.29
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (1R,2R)-1,2-Bis(2-hydroxyphenyl)ethylenediamine