missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1R,2R)-1,2-Bis(2,4,6-trimethylphenyl)ethylenediamine 97.0+%, TCI America™
$365.35 - $1252.65
Chemical Identifiers
| CAS | 425615-42-5 |
|---|---|
| Molecular Formula | C20H28N2 |
| Molecular Weight (g/mol) | 296.458 |
| MDL Number | MFCD06797063 |
| InChI Key | ILMRHFMYIXTNMC-WOJBJXKFSA-N |
| Synonym | (1R,2R)-1,2-Diamino-1,2-dimesitylethane, (1R,2R)-1,2-Dimesitylethylenediamine, (1R,2R)-1,2-Dimesitylethane-1,2-diamine |
| PubChem CID | 29963716 |
| IUPAC Name | (1R,2R)-1,2-bis(2,4,6-trimethylphenyl)ethane-1,2-diamine |
| SMILES | CC1=CC(=C(C(=C1)C)C(C(C2=C(C=C(C=C2C)C)C)N)N)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B2316100MG
|
TCI America
B2316100MG |
100 mg |
Each for $365.35
|
|
|||||
|
B2316500MG
|
TCI America
B2316500MG |
500 mg |
Each for $1,252.65
|
|
|||||
Chemical Identifiers
| 425615-42-5 | |
| 296.458 | |
| ILMRHFMYIXTNMC-WOJBJXKFSA-N | |
| 29963716 | |
| CC1=CC(=C(C(=C1)C)C(C(C2=C(C=C(C=C2C)C)C)N)N)C |
| C20H28N2 | |
| MFCD06797063 | |
| (1R,2R)-1,2-Diamino-1,2-dimesitylethane, (1R,2R)-1,2-Dimesitylethylenediamine, (1R,2R)-1,2-Dimesitylethane-1,2-diamine | |
| (1R,2R)-1,2-bis(2,4,6-trimethylphenyl)ethane-1,2-diamine |
Specifications
| 425615-42-5 | |
| White-Yellow | |
| MFCD06797063 | |
| (1R,2R)-1,2-Diamino-1,2-dimesitylethane, (1R,2R)-1,2-Dimesitylethylenediamine, (1R,2R)-1,2-Dimesitylethane-1,2-diamine | |
| CC1=CC(=C(C(=C1)C)C(C(C2=C(C=C(C=C2C)C)C)N)N)C | |
| 296.458 | |
| 296.46 | |
| Crystalline Powder |
| 86°C | |
| C20H28N2 | |
| 100 mg | |
| ILMRHFMYIXTNMC-WOJBJXKFSA-N | |
| (1R,2R)-1,2-bis(2,4,6-trimethylphenyl)ethane-1,2-diamine | |
| 29963716 | |
| ≥97.0% (GC) | |
| (1R,2R)-1,2-Bis(2,4,6-trimethylphenyl)ethylenediamine |
Safety and Handling
TSCA : No