missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R)-1,2-Bis(2,4,6-trimethylphenyl)ethylenediamine 97.0+%, TCI America™

Catalog No. b2316500mg
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2316500MG 500 mg
B2316100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B2316500MG Supplier TCI America Supplier No. B2316500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 425615-42-5
Molecular Formula C20H28N2
Molecular Weight (g/mol) 296.458
MDL Number MFCD06797063
InChI Key ILMRHFMYIXTNMC-WOJBJXKFSA-N
Synonym (1R,2R)-1,2-Diamino-1,2-dimesitylethane, (1R,2R)-1,2-Dimesitylethylenediamine, (1R,2R)-1,2-Dimesitylethane-1,2-diamine
PubChem CID 29963716
IUPAC Name (1R,2R)-1,2-bis(2,4,6-trimethylphenyl)ethane-1,2-diamine
SMILES CC1=CC(=C(C(=C1)C)C(C(C2=C(C=C(C=C2C)C)C)N)N)C

Specifications

Melting Point 86°C
Color White-Yellow
Quantity 500 mg
Formula Weight 296.46
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (1R,2R)-1,2-Bis(2,4,6-trimethylphenyl)ethylenediamine