Learn More
Thermo Scientific™ 1M TEAB and 50% Hydroxylamine Buffers for TMT Experiments
Catalog No. PI90115
Change view
Click to view available options
Product Type:
Hydroxylamine
Triethylammonium Bicarbonate (TEAB)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
PI90115 Hydroxylamine
PI90114 Triethylammonium Bicarbonate (TEAB)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI90115 Supplier Thermo Scientific™ Supplier No. 90115
Season of Savings
View all promotions
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Common TMT buffers such as 1M Triethylammonium bicarbonate (TEAB) and 50% Hydroxylamine for isobaric mass tag labeling experiments.
Pre-formulated TEAB and 50% hydroxylamine designed for use in TMT labeling and quenching of peptides.
Triethylammonium bicarbonate (TEAB) is a 10X dissolution buffer for isobaric mass tag labeling experiments. 50% hydroxylamine is a 10X quenching reagent for amine-reactive labeling for TMT experiments.
Specifications
Product Type Hydroxylamine
Product Line TMT
Content And Storage Store protected from direct sunlight in a dry, cool, and well-ventilated area.
Starting Material Cell Lysate
Workflow Step Protein Labeling
Quantity 5 mL