missing translation for 'onlineSavingsMsg'
Learn More

1M KNO3 Electrolyte Solution, METTLER TOLEDO™

Catalog No. 01912585
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01912585 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01912585 Supplier METTLER TOLEDO Supplier No. 51350078
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For optimum performance of Ion-Selective Electrode (ISE) Half-Cells

  • 1M KNO3 Electrolyte
  • For optimum performance of Ion-Selective Electrode (ISE) half-cells

Specifications

Chemical Name or Material 1 M KNO3 Electrolyte
Color Colorless
CAS 7757-79-1
Boiling Point 400°C
Melting Point 334°C
Molecular Formula KNO3
Synonym Nitric Acid, Potassium Salt, Saltpeter
Molecular Weight (g/mol) 101.1
Packaging Plastic Bottle
pH No Information Available
Quantity 250 mL
Show More Show Less