missing translation for 'onlineSavingsMsg'
Learn More

1H-Tridecafluorohexane 97.0+%, TCI America™

Catalog No. T24965G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T24965G Supplier TCI America Supplier No. T24965G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 355-37-3
Molecular Formula C6HF13
Molecular Weight (g/mol) 320.05
MDL Number MFCD00142632
InChI Key XJSRKJAHJGCPGC-UHFFFAOYSA-N
Synonym 1h-perfluorohexane, 1h-tridecafluorohexane, hexane, tridecafluoro, trideca-1,1,1,2,2,3,3,4,4,5,5,6,6-fluorohexane, hexane, 1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro, tridecafluorohexane, acmc-209tqg, #
PubChem CID 67730
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluorohexane
SMILES FC(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Color Yellow
Boiling Point 70°C
Quantity 5 g
Formula Weight 320.05
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1H-Tridecafluorohexane
EINECSNumber (2)-3771
TSCA No