CAS RN 355-37-3
IUPAC Name:
1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluorohexane
Synonyms:
1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluorohexane
Molecular Weight (g/mol):
320.05
Molecular Formula:
C6HF13
InChi Key:
XJSRKJAHJGCPGC-UHFFFAOYSA-N
SMILES:
FC(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
1
–
1
of
1
results
1
1H-Tridecafluorohexane 97.0+%, TCI America™
CAS: 355-37-3 Molecular Formula: C6HF13 Molecular Weight (g/mol): 320.05 MDL Number: MFCD00142632 InChI Key: XJSRKJAHJGCPGC-UHFFFAOYSA-N Synonym: 1h-perfluorohexane,1h-tridecafluorohexane,hexane, tridecafluoro,trideca-1,1,1,2,2,3,3,4,4,5,5,6,6-fluorohexane,hexane, 1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro,tridecafluorohexane,acmc-209tqg,# PubChem CID: 67730 IUPAC Name: 1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluorohexane SMILES: FC(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F