missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1H-Tetrazole-5-acetic Acid 98.0+%, TCI America™
Supplier: TCI America T273725G
Specifications
| 1H-Tetrazole-5-acetic Acid | |
| 177°C | |
| C3H4N4O2 | |
| 25 g | |
| JUNAPQMUUHSYOV-UHFFFAOYSA-N | |
| 2-(2H-tetrazol-5-yl)acetic acid | |
| 89031 | |
| ≥98.0% (GC,T) |
| 21743-75-9 | |
| White | |
| MFCD03094010 | |
| 1325 | |
| C(C1=NNN=N1)C(=O)O | |
| 128.091 | |
| 128.09 | |
| Crystalline Powder |
Chemical Identifiers
| 21743-75-9 | |
| 128.091 | |
| JUNAPQMUUHSYOV-UHFFFAOYSA-N | |
| 2-(2H-tetrazol-5-yl)acetic acid |
| C3H4N4O2 | |
| MFCD03094010 | |
| 89031 | |
| C(C1=NNN=N1)C(=O)O |
Safety and Handling
TSCA : No