missing translation for 'onlineSavingsMsg'
Learn More

1H-Tetrazole, 0.45M in acetonitrile

Catalog No. aaj61034ah
Change view
Click to view available options
Quantity:
200 mL
450 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61034AH 200 mL
AAJ61034AO 450 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61034AH Supplier Thermo Scientific Chemicals Supplier No. J61034AH

Coupling reagent for preparation of polynucleotides | CAS: 288-94-8 | CH2N4 | 70.055 g/mol

1H-Tetrazole is used as a bioisostere for the carboxylate group. It is also used as coupling reagent for preparation of polynucleotides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1H-Tetrazole is used as a bioisostere for the carboxylate group. It is also used as coupling reagent for preparation of polynucleotides.

Solubility
Miscible with water.

Notes
Incompatible with reducing agents and alkali metals.

Chemical Identifiers

CAS 288-94-8
Molecular Formula CH2N4
Molecular Weight (g/mol) 70.055
MDL Number MFCD00005247
InChI Key KJUGUADJHNHALS-UHFFFAOYSA-N
PubChem CID 67519
ChEBI CHEBI:33193
IUPAC Name 2H-tetrazole
SMILES C1=NNN=N1

Specifications

Density 0.798
Quantity 200 mL
UN Number UN1648
Beilstein 105799
Merck Index 14,2786
Synonym 1h-tetrazole, tetrazole, tetraazacyclopentadiene, 1-h-tetrazole, 1h-1,2,3,4-tetrazole, 1 h-tetrazole, 1h-tetrazole solution, 2h-1,2,3,4-tetrazole, racemic tetrazole, 1h-tetraazole
Solubility Information Miscible with water.
Formula Weight 70.05
Concentration or Composition (by Analyte or Components) 0.45M in acetonitrile
Physical Form Liquid
Chemical Name or Material 1H-Tetrazole
Show More Show Less
Hazard Category H225-H302+H312+H332-H319
Hazard Statement GHS H Statement
H225-H312-H332-H319
Highly flammable liquid and vapor.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ACETONITRILE
EINECSNumber 206-023-4
RTECSNumber UW7370000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only