accessibility menu, dialog, popup

UserName

HNNNN

CAS RN 288-94-8

IUPAC Name: 2H-1,2,3,4-tetrazole
Synonyms: 2H-tetrazole
Molecular Weight (g/mol): 70.06
Molecular Formula: CH2N4
InChi Key: KJUGUADJHNHALS-UHFFFAOYSA-N
SMILES: N1N=CN=N1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)