missing translation for 'onlineSavingsMsg'
Learn More

1H-Pyrrolo[3,2-c]pyridine 98.0+%, TCI America™

Catalog No. P1896100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1896100MG 100 mg
P18961G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P1896100MG Supplier TCI America Supplier No. P1896100MG

Chemical Identifiers

CAS 271-34-1
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.139
MDL Number MFCD00955936
InChI Key SRSKXJVMVSSSHB-UHFFFAOYSA-N
Synonym 5-azaindole, 1h-pyrrolo 3,2-c pyridine, 1,5-diazaindene, 3,7-dideazapurine, 5-azindole, 5h-pyrrolo 3,2-c pyridine, pyrrolo 3,2-c pyridine, 5-aza indole, 5-aza-indole
PubChem CID 9220
IUPAC Name 1H-pyrrolo[3,2-c]pyridine
SMILES C1=CNC2=C1C=NC=C2

Specifications

Melting Point 113°C
Color White-Yellow
Boiling Point 132°C
Quantity 100 mg
Formula Weight 118.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1H-Pyrrolo[3,2-c]pyridine