missing translation for 'onlineSavingsMsg'
Learn More

1H-Pyrrolo[3,2-b]pyridine 98.0+%, TCI America™

Catalog No. p-7135372
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P20411G 1 g
P20415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P20411G Supplier TCI America Supplier No. P20411G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 272-49-1
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.14
MDL Number MFCD00971977
InChI Key XWIYUCRMWCHYJR-UHFFFAOYSA-N
Synonym 4-azaindole, 1h-pyrrolo 3,2-b pyridine, 1,4-diazaindene, pyrrolopyridine, 4h-pyrrolo 3,2-b pyridine, 1h-pyrrolo 3.2-b pyridine, zlchem 17, pubchem7675, acmc-1clkx
PubChem CID 9226
IUPAC Name 1H-pyrrolo[3,2-b]pyridine
SMILES N1C=CC2=NC=CC=C12

Specifications

Melting Point 128°C
Color Yellow
Quantity 1 g
Formula Weight 118.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1H-Pyrrolo[3,2-b]pyridine
RTECSNumber UY8715000
TSCA No