missing translation for 'onlineSavingsMsg'
Learn More

1H-Pyrazole-3-boronic acid hydrate, 95%

Catalog No. AAH2728603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2728603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2728603 Supplier Thermo Scientific Chemicals Supplier No. H2728603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 376584-63-3 | C3H5BN2O2 | 111.90 g/mol

Significantly used as synthetic intermediates. They are widely used in the production of optics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Significantly used as synthetic intermediates. They are widely used in the production of optics.

Solubility
Soluble in water.

Notes
Refrigerated (2-8°C). Moisture sensitive. Store under inert gas. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 376584-63-3
Molecular Formula C3H5BN2O2
Molecular Weight (g/mol) 111.90
MDL Number MFCD07368247 MFCD02020768
InChI Key NEUWPDLMDVINSN-UHFFFAOYSA-N
Synonym 1h-pyrazol-3-yl boronic acid, pyrazole-3-boronic acid, 1h-pyrazol-3-ylboronic acid, 1h-pyrazole-3-boronic acid, 1h-pyrazole-5-boronic acid, 1h-pyrazol-5-yl boronic acid, 1h-pyrazol-5-yl-5-boronic acid, pyrazol-3-ylboronic acid, 2h-pyrazol-3-ylboronic acid
PubChem CID 11251979
IUPAC Name 1H-pyrazol-5-ylboronic acid
SMILES OB(O)C1=NNC=C1

Specifications

Melting Point 85°C to 88°C
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 111.9
Percent Purity 95%
Chemical Name or Material 1H-Pyrazole-3-boronic acid hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only