accessibility menu, dialog, popup

UserName

CAS RN 376584-63-3

IUPAC Name: (1H-pyrazol-3-yl)boronic acid
Synonyms: 1H-pyrazol-3-ylboronic acid
Molecular Weight (g/mol): 111.9
Molecular Formula: C3H5BN2O2
InChi Key: NEUWPDLMDVINSN-UHFFFAOYSA-N
SMILES: OB(O)C1=NNC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results