Learn More
1H-Pyrazole-1-carboxamidine hydrochloride, 99%
CAS: 4023-02-3 | C4H6N4 | 110.12 g/mol
$107.68 - $372.48
Chemical Identifiers
| CAS | 4023-02-3 |
|---|---|
| MDL Number | MFCD00210087 |
| IUPAC Name | pyrazole-1-carboximidamide;hydrochloride |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6063109
|
Thermo Scientific Chemicals
H6063109 |
10 g |
Each for $107.68
|
|
|||||
|
AAH6063118
|
Thermo Scientific Chemicals
H6063118 |
50 g |
Each for $372.48
|
|
|||||
Description
1H-Pyrazole-1-carboxamidine hydrochloride may be used in the following studies:
- Preparation of guanidylated hollow fiber membranes.
- SGuanylation of amines and in peptide synthesis.
- Synthesis of bis-guanidinium-cholesterol derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1H-Pyrazole-1-carboxamidine hydrochloride may be used in the following studies: • Preparation of guanidylated hollow fiber membranes. • Guanylation of amines and in peptide synthesis. • Synthesis of bis-guanidinium-cholesterol derivatives.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents. Store under room temperature.
Chemical Identifiers
| 4023-02-3 | |
| MFCD00210087 | |
| pyrazole-1-carboximidamide;hydrochloride |
Specifications
| 4023-02-3 | |
| C12H5ClO3 | |
| 10 g | |
| Soluble in water. | |
| C1=CC2=C(C=CC3=C2C(=C1)C(=O)OC3=O)Cl | |
| 77672 | |
| 99% |
| 167°C to 170°C | |
| MFCD00210087 | |
| 4-chloro-1,8-naphthalic anhydride, 4-chloronaphthalic anhydride, 6-chlorobenzo de isochromene-1,3-dione, 4-chloronapthalic anhydride, 6-chloro-1h,3h-benzo de isochromene-1,3-dione, 4-chloronaphthalene-1,8-dicarboxylic anhydride, 1h,3h-naphtho 1,8-cd pyran-1,3-dione, 6-chloro, naphthalic anhydride, 4-chloro-7ci,8ci, 6-chloro-1h,3h-naphtho 1,8-cd pyran-1,3-dione | |
| UJEUBSWHCGDJQU-UHFFFAOYSA-N | |
| 232.62 | |
| 146.58 | |
| 1H-Pyrazole-1-carboxamidine hydrochloride |
Safety and Handling
GHS H Statement
H318-H373-H302-H317
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
May cause an allergic skin reaction.
P260-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P310-P314-P330-P333+P313-P363-P501c
H302-H317-H318-H373
EINECSNumber : 429-520-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only