missing translation for 'onlineSavingsMsg'
Learn More

1H-Pyrazole-1-carboxamidine hydrochloride, 99%

Catalog No. AAH6063118
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6063118 50 g
AAH6063109 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6063118 Supplier Thermo Scientific Chemicals Supplier No. H6063118
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4023-02-3 | C4H6N4 | 110.12 g/mol

1H-Pyrazole-1-carboxamidine hydrochloride may be used in the following studies:

  • Preparation of guanidylated hollow fiber membranes.
  • SGuanylation of amines and in peptide synthesis.
  • Synthesis of bis-guanidinium-cholesterol derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1H-Pyrazole-1-carboxamidine hydrochloride may be used in the following studies: • Preparation of guanidylated hollow fiber membranes. • Guanylation of amines and in peptide synthesis. • Synthesis of bis-guanidinium-cholesterol derivatives.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents. Store under room temperature.

Chemical Identifiers

CAS 4023-02-3
MDL Number MFCD00210087
IUPAC Name pyrazole-1-carboximidamide;hydrochloride

Specifications

Melting Point 167°C to 170°C
Molecular Formula C4H6N4
Quantity 50 g
Synonym 1h-pyrazole-1-carboxamidine hydrochloride, 1h-pyrazole-1-carboximidamide hydrochloride, 1-amidinopyrazole hydrochloride, praxadine, pyrazole-1-carboxamidine hydrochloride, pyrazole-1-carboximidamide hydrochloride, pyrazole-1-carboxamidine hcl, 1h-pyrazole-1-carboxamidinehydrochloride, praxadine hydrochloride, 1h-pyrazole-1-carboxamidine hcl
Solubility Information Soluble in water.
InChI Key UCQFSGCWHRTMGG-UHFFFAOYSA-N
SMILES C1=CN(N=C1)C(=N)N.Cl
Molecular Weight (g/mol) 110.12
PubChem CID 2734672
Formula Weight 146.58
Percent Purity 99%
Chemical Name or Material 1H-Pyrazole-1-carboxamidine hydrochloride
Show More Show Less
Hazard Category H302-H317-H318-H373
Hazard Statement GHS H Statement
H318-H373-H302-H317
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P260-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P310-P314-P330-P333+P313-P363-P501c
EINECSNumber 429-520-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only