missing translation for 'onlineSavingsMsg'
Learn More

1H-Indole-7-carbohydrazide, 97%, Thermo Scientific™

Catalog No. SB02108CB
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SB02108CB 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SB02108CB Supplier Thermo Scientific Chemicals Supplier No. SB02108CB

Chemical Identifiers

CAS 321309-24-4
Molecular Formula C9H9N3O
Molecular Weight (g/mol) 175.191
MDL Number MFCD00572868
InChI Key NCFMBDUFIJHHAW-UHFFFAOYSA-N
Synonym 1h-indole-7-carboxylic acid hydrazide, 7-hydrazinocarbonyl-1h-indole, 1h-indole-7-carboxylic acid, hydrazide, indole-7-carbohydrazide, 1h-indole-7-carboxylicacid, hydrazide
PubChem CID 712457
IUPAC Name 1H-indole-7-carbohydrazide
SMILES C1=CC2=C(C(=C1)C(=O)NN)NC=C2

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 175.19
Percent Purity 97%
Chemical Name or Material 1H-indole-7-carbohydrazide
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.