missing translation for 'onlineSavingsMsg'
Learn More

1H-Indole-2-methanol, 96%

Catalog No. AC459810010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC459810010 1 g
AC459810050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC459810010 Supplier Thermo Scientific Chemicals Supplier No. 459810010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 24621-70-3 | C9H9NO | 147.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 24621-70-3
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.17
InChI Key XEEANGGQJOWRTG-UHFFFAOYSA-N
Synonym 1h-indol-2-yl methanol, 1h-indole-2-methanol, 1h-indol-2-yl-methanol, 2-hydroxymethylindole, 2-hydroxymethyl indole, hydroxymethylindol, 2-indolemethanol, indole-2-methanol, indol-2-ylmethan-1-ol
PubChem CID 98783
IUPAC Name 1H-indol-2-ylmethanol
SMILES C1=CC=C2C(=C1)C=C(N2)CO

Specifications

Melting Point 72.0°C to 78.0°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 147.17
Percent Purity 96%
Chemical Name or Material 1H-Indole-2-methanol
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

RUO – Research Use Only