CAS RN 24621-70-3
IUPAC Name:
(1H-indol-2-yl)methanol
Synonyms:
1H-indol-2-ylmethanol
Molecular Weight (g/mol):
147.18
Molecular Formula:
C9H9NO
InChi Key:
XEEANGGQJOWRTG-UHFFFAOYSA-N
SMILES:
OCC1=CC2=CC=CC=C2N1
1
–
1
of
1
results
1
Promotions Available
1H-Indole-2-methanol, 96%
1H-Indole-2-methanol, 96%
CAS: 24621-70-3 Molecular Formula: C9H9NO Molecular Weight (g/mol): 147.17 InChI Key: XEEANGGQJOWRTG-UHFFFAOYSA-N Synonym: 1h-indol-2-yl methanol,1h-indole-2-methanol,1h-indol-2-yl-methanol,2-hydroxymethylindole,2-hydroxymethyl indole,hydroxymethylindol,2-indolemethanol,indole-2-methanol,indol-2-ylmethan-1-ol PubChem CID: 98783 IUPAC Name: 1H-indol-2-ylmethanol SMILES: C1=CC=C2C(=C1)C=C(N2)CO