missing translation for 'onlineSavingsMsg'
Learn More

1H-Indole-2-carboxylic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 1477-50-5
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00005611
InChI Key HCUARRIEZVDMPT-UHFFFAOYSA-N
Synonym indole-2-carboxylic acid, 2-carboxyindole, 2-indolecarboxylic acid, indol-2-carboxylic acid, 1h-indolecarboxylic acid, indole-2-carboxylate, 2-indolylformic acid, 2-indole carboxylic acid, 1h-indol-2-carbons, carboxyindole
PubChem CID 72899
IUPAC Name 1H-indole-2-carboxylic acid
SMILES OC(=O)C1=CC2=CC=CC=C2N1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
CC12801DA
View Documents
N/A 1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

1477-50-5
161.16
HCUARRIEZVDMPT-UHFFFAOYSA-N
72899
OC(=O)C1=CC2=CC=CC=C2N1
C9H7NO2
MFCD00005611
indole-2-carboxylic acid, 2-carboxyindole, 2-indolecarboxylic acid, indol-2-carboxylic acid, 1h-indolecarboxylic acid, indole-2-carboxylate, 2-indolylformic acid, 2-indole carboxylic acid, 1h-indol-2-carbons, carboxyindole
1H-indole-2-carboxylic acid

Specifications

1477-50-5
Amber glass bottle
MFCD00005611
HCUARRIEZVDMPT-UHFFFAOYSA-N
1H-indole-2-carboxylic acid
72899
97%
97%
C9H7NO2
indole-2-carboxylic acid, 2-carboxyindole, 2-indolecarboxylic acid, indol-2-carboxylic acid, 1h-indolecarboxylic acid, indole-2-carboxylate, 2-indolylformic acid, 2-indole carboxylic acid, 1h-indol-2-carbons, carboxyindole
OC(=O)C1=CC2=CC=CC=C2N1
161.16
161.16
1H-Indole-2-carboxylic acid
Videos
Safety and Handling

Safety and Handling

Warning

SDS
Documents

Documents