missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1H-Indole-2-carboxylic acid, 97%, Thermo Scientific™
Specifications
Specifications
| Assay Percent Range | 97% |
| Packaging | Amber glass bottle |
| Quantity | 1g |
| Formula Weight | 161.16 |
| Percent Purity | 97% |
| Chemical Name or Material | 1H-Indole-2-carboxylic acid |
Safety and Handling
| Hazard Category | Warning |