Learn More
1H-Benzotriazole, 99%
CAS: 95-14-7 | C6H5N3 | 119.127 g/mol
Supplier: Thermo Scientific Chemicals A154230B
Description
1H-Benzotriazole is an anticorrosive agent, which is useful in aircraft deicing and antifreeze fluids. It is also employed in dishwasher detergents. Further, it is used as a restrainer in photographic emulsions and also useful as a reagent for the determination of silver in analytical chemistry. It also serves as a corrosion inhibitor in the atmosphere and underwater. Further, it is utilized in the synthesis of amines from glyoxal.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1H-Benzotriazole is an anticorrosive agent, which is useful in aircraft deicing and antifreeze fluids. It is also employed in dishwasher detergents. Further, it is used as a restrainer in photographic emulsions and also useful as a reagent for the determination of silver in analytical chemistry. It also serves as a corrosion inhibitor in the atmosphere and underwater. Further, it is utilized in the synthesis of amines from glyoxal.
Solubility
Soluble in water, benzene, toluene, chloroform, ethanol and N,N-dimethylformamide.
Notes
Store in a cool place. Incompatible with strong oxidizing agents and heavy metals.
Specifications
| 1H-Benzotriazole | |
| 96°C to 99°C | |
| 6.0 to 7.0 | |
| 212°C (413°F) | |
| Odorless | |
| MFCD00005699 | |
| 112133 | |
| 1h-benzotriazole, benzotriazole, 1,2,3-benzotriazole, 1h-benzo d 1,2,3 triazole, azimidobenzene, 1h-1,2,3-benzotriazole, aziminobenzene, benztriazole, benzene azimide, benzisotriazole | |
| QRUDEWIWKLJBPS-UHFFFAOYSA-N | |
| 2H-benzotriazole | |
| 7220 | |
| 119.13 |
| 95-14-7 | |
| 1.36 | |
| 204°C (15 mmHg) | |
| 99% | |
| C6H5N3 | |
| 1000 g | |
| 14,1108 | |
| Soluble in water,benzene,toluene,chloroform,ethanol and N,N-dimethylformamide. | |
| C1=CC2=NNN=C2C=C1 | |
| 119.127 | |
| CHEBI:75331 | |
| 99% |
Chemical Identifiers
| 95-14-7 | |
| 119.127 | |
| QRUDEWIWKLJBPS-UHFFFAOYSA-N | |
| 7220 | |
| 2H-benzotriazole |
| C6H5N3 | |
| MFCD00005699 | |
| 1h-benzotriazole, benzotriazole, 1,2,3-benzotriazole, 1h-benzo d 1,2,3 triazole, azimidobenzene, 1h-1,2,3-benzotriazole, aziminobenzene, benztriazole, benzene azimide, benzisotriazole | |
| CHEBI:75331 | |
| C1=CC2=NNN=C2C=C1 |
Safety and Handling
GHS H Statement
H302-H332-H319
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
H302-H319
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 202-394-1
RTECSNumber : DM1225000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only