missing translation for 'onlineSavingsMsg'
Learn More

1H-Benzotriazole, 99%

Catalog No. p-7022579
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1542318 50 g
AAA1542330 250 g
AAA154230B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1542318 Supplier Thermo Scientific Chemicals Supplier No. A1542318
Only null left

CAS: 95-14-7 | C6H5N3 | 119.127 g/mol

1H-Benzotriazole is an anticorrosive agent, which is useful in aircraft deicing and antifreeze fluids. It is also employed in dishwasher detergents. Further, it is used as a restrainer in photographic emulsions and also useful as a reagent for the determination of silver in analytical chemistry. It also serves as a corrosion inhibitor in the atmosphere and underwater. Further, it is utilized in the synthesis of amines from glyoxal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1H-Benzotriazole is an anticorrosive agent, which is useful in aircraft deicing and antifreeze fluids. It is also employed in dishwasher detergents. Further, it is used as a restrainer in photographic emulsions and also useful as a reagent for the determination of silver in analytical chemistry. It also serves as a corrosion inhibitor in the atmosphere and underwater. Further, it is utilized in the synthesis of amines from glyoxal.

Solubility
Soluble in water, benzene, toluene, chloroform, ethanol and N,N-dimethylformamide.

Notes
Store in a cool place. Incompatible with strong oxidizing agents and heavy metals.

Chemical Identifiers

CAS 95-14-7
Molecular Formula C6H5N3
Molecular Weight (g/mol) 119.127
MDL Number MFCD00005699
InChI Key QRUDEWIWKLJBPS-UHFFFAOYSA-N
Synonym 1h-benzotriazole, benzotriazole, 1,2,3-benzotriazole, 1h-benzo d 1,2,3 triazole, azimidobenzene, 1h-1,2,3-benzotriazole, aziminobenzene, benztriazole, benzene azimide, benzisotriazole
PubChem CID 7220
ChEBI CHEBI:75331
IUPAC Name 2H-benzotriazole
SMILES C1=CC2=NNN=C2C=C1

Specifications

Melting Point 96°C to 99°C
Density 1.360
Boiling Point 204°C (15 mm)
Flash Point 212°C (413°F)
Assay Percent Range 99%
Quantity 50 g
Beilstein 112133
Merck Index 14,1108
Solubility Information Soluble in water,benzene,toluene,chloroform,ethanol and N,N-dimethylformamide.
Formula Weight 119.13
Percent Purity 99%
Chemical Name or Material Benzotriazole
Show More Show Less
Hazard Category H302-H319
Hazard Statement GHS H Statement
H302-H332-H319
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 202-394-1
RTECSNumber DM1225000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only