missing translation for 'onlineSavingsMsg'
Learn More

1H-Benzotriazol-1-yloxytris(dimethylamino)phosphonium hexafluorophosphate, 98%

Catalog No. AAA1614006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1614006 5 g
AAA1614014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1614006 Supplier Thermo Scientific Chemicals Supplier No. A1614006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 56602-33-6 | C12H22F6N6OP2 | 442.287 g/mol

1H-Benzotriazol-1-yloxytris(dimethylamino)phosphonium hexafluorophosphate is used as a reagent for peptide coupling, lactonization, selective esterification, amidation of alfa amino acids without racemization and synthesis of magnolamide for antioxidative activity and catalyst for 9-acridinecaroboxamide derivative. It is also used as a precursor for the synthesis of phenyl esters of amino acids. It acts as a substitute for (Benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (BOP) reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1H-Benzotriazol-1-yloxytris(dimethylamino)phosphonium hexafluorophosphate is used as a reagent for peptide coupling, lactonization, selective esterification, amidation of alfa amino acids without racemization and synthesis of magnolamide for antioxidative activity and catalyst for 9-acridinecaroboxamide derivative. It is also used as a precursor for the synthesis of phenyl esters of amino acids. It acts as a substitute for (Benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (BOP) reagent.

Solubility
Soluble in methanol, acetone and dichloromethane. Insoluble in water.

Notes
Moisture and light sensitive. Incompatible with strong oxidizing agents. Store in a cool place. Keep the container tightly closed.

Chemical Identifiers

CAS 56602-33-6
Molecular Formula C12H22F6N6OP2
Molecular Weight (g/mol) 442.287
MDL Number MFCD00011948
InChI Key MGEVGECQZUIPSV-UHFFFAOYSA-N
Synonym bop reagent, 1h-benzo d 1,2,3 triazol-1-yl oxy tris dimethylamino phosphonium hexafluorophosphate v, castro's reagent, bop-reagent, benzotriazol-1-yloxytris dimethylamino-phosphonium hexafluorophosphate, ccris 2602, castros reagent, tri dimethylamino benzotriazol-1-yloxyphosphonium hexafluorophosphate, benzotriazol-1-yloxy tris dimethylamino phosphonium hexafluorophosphate, 1h-benzotriazol-1-yloxytris dimethylamino phosphonium hexafluorophosphate
PubChem CID 151348
IUPAC Name benzotriazol-1-yloxy-tris(dimethylamino)phosphanium;hexafluorophosphate
SMILES CN(C)[P+](N(C)C)(N(C)C)ON1C2=CC=CC=C2N=N1.F[P-](F)(F)(F)(F)F

Specifications

Melting Point ∼138°C (decomposition)
Quantity 5 g
UN Number UN1325
Beilstein 4287025
Sensitivity Light and moisture sensitive
Solubility Information Soluble in methanol,acetone and dichloromethane. Insoluble in water.
Formula Weight 442.29
Percent Purity 98%
Chemical Name or Material 1H-Benzotriazol-1-yloxytris(dimethylamino)phosphonium hexafluorophosphate
Hazard Category H228-H315-H319-H335-H500
Hazard Statement GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
EINECSNumber 260-279-1
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only